Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNASE3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140984
This item is not returnable.
View return policy
Description
Application notes: Indirect ELISA.
ECP (eosinophil cationic protein), also known as RNS3 or RNASE3 (ribonuclease, RNase A family, 3), is a 160 amino acid potent cytotoxic secretory protein that belongs to the pancreatic ribonuclease family. ECP localizes to the granule matrix of the eosinophil and is involved in the immune response system. ECP is a cytotoxin, neurotoxin and helminthotoxin that is secreted by activated human eosinophils. ECP exhibits anti-parasitic, antibacterial and ribonucleolytic activities. It has been suggested that ECP induces the neurotoxic effect known as the Gordon phenomenon, a syndrome manifested by ataxia, muscular rigidity, paralysis and tremor that may lead to death. ECP is considered a marker of eosinophilic inflammation and high levels have been found in cases of active asthma and other allergic diseases.
Specifications
RNASE3 | |
Polyclonal | |
Unconjugated | |
RNASE3 | |
cytotoxic ribonuclease; Ear11; EAR-11; Ear3; ECP; Eosinophil cationic protein; eosinophil cationic protein-like; eosinophil cationic-type ribonuclease 3; Eosinophil secondary granule ribonuclease 11; eosinophil-associated ribonuclease 3; eosinophil-associated, ribonuclease A family, member 3; LOC100361866; mR3; MR-3; murine ribonuclease 3; RAF1; Ribonuclease 3; ribonuclease A family member 3; ribonuclease, RNase A family 3; ribonuclease, RNase A family, 3; RNase 3; RNASE3; RNS3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6037 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P12724 | |
RNASE3 | |
recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction