Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNASE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595960 Supplier Invitrogen™ Supplier No. PA595960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPPQFTRAQW FAIQHISLNP PRCTIAMRAI NNYRWRCKNQ NTFLRTTFAN VVNVCGNQSI RCPHNRTLNN CHRSRFRVPL LHCDLINPGA QNISNCTYAD RPGRRFYVVA CDNRDPRDSP RYPVVPVHLD TTI.

ECP (eosinophil cationic protein), also known as RNS3 or RNASE3 (ribonuclease, RNase A family, 3), is a 160 amino acid potent cytotoxic secretory protein that belongs to the pancreatic ribonuclease family. ECP localizes to the granule matrix of the eosinophil and is involved in the immune response system. ECP is a cytotoxin, neurotoxin and helminthotoxin that is secreted by activated human eosinophils. ECP exhibits anti-parasitic, antibacterial and ribonucleolytic activities. It has been suggested that ECP induces the neurotoxic effect known as the Gordon phenomenon, a syndrome manifested by ataxia, muscular rigidity, paralysis and tremor that may lead to death. ECP is considered a marker of eosinophilic inflammation and high levels have been found in cases of active asthma and other allergic diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNASE3
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNASE3
Gene Accession No. P12724
Gene Alias cytotoxic ribonuclease; Ear11; EAR-11; Ear3; ECP; Eosinophil cationic protein; eosinophil cationic protein-like; eosinophil cationic-type ribonuclease 3; Eosinophil secondary granule ribonuclease 11; eosinophil-associated ribonuclease 3; eosinophil-associated, ribonuclease A family, member 3; LOC100361866; mR3; MR-3; murine ribonuclease 3; RAF1; Ribonuclease 3; ribonuclease A family member 3; ribonuclease, RNase A family 3; ribonuclease, RNase A family, 3; RNase 3; RNASE3; RNS3
Gene Symbols RNASE3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 28-160 of human RNASE3 (NP_0029262).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6037
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.