Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNASE4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5121641
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121641 20 μL
PIPA5121640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121641 Supplier Invitrogen™ Supplier No. PA5121641
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNase 4 and RNase 5/ang 1 are unique among the RNase A ribonuclease genes in that they maintain a complex gene locus that is conserved across species with transcription initiated from tissue-specific dual promoters followed by differential exon splicing. Rnase4 (ribonuclease, RNase A family 4) gene can produce 2 transcripts both encoding 148 amino acid proteins. Rnase 4 is a member of the pancreatic-type of secretory ribonucleases, a subset of the ribonuclease A superfamily. RNase 4 prefers poly(C) as a substrate and hydrolyzes 2',3'-cyclic nucleotides, with a pH optimum near 8.0. mRNA encoding RNase 4 is detectable in human pancreas, lung, skeletal muscle, heart, kidney and placenta; liver represents the most abundant source.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNASE4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene RNASE4
Gene Accession No. O55004, P34096
Gene Alias C730049F20Rik; Rab1; Ribonuclease 4; ribonuclease A B1; ribonuclease A family member 4; ribonuclease, RNase A family 4; ribonuclease, RNase A family, 4; RL3; RNase 4; RNASE4; RNS4
Gene Symbols RNASE4
Host Species Rabbit
Immunogen Recombinant RNASE4 (Asp30-Gly147) expressed in E.coli.
Purification Method Protein A, Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56759, 6038
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.