Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNASE6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551442
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551442 Supplier Invitrogen™ Supplier No. PA551442
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKRITKAHWF EIQHIQPSPL QCNRAMSGIN NYTQHCKHQN TFLHDSFQNV AAVCDLLSIV CKNRRHNCHQ SSKPVNMTDC RLTSGKYPQC RYSAAAQYKF FIVACDPPQK SDPPYKLVPV Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 66%.

RNASE6 may have a role in host defense.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNASE6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNASE6
Gene Accession No. Q93091
Gene Alias 9530043P15Rik; BB119466; RAD1; ribonuclease A D1; ribonuclease A family member k6; ribonuclease A family, member 6; ribonuclease K6; ribonuclease, RNase A family, 6; ribonuclease, RNase A family, k6; RNase K6; RNASE6; RNasek6; RNS6; testicular tissue protein Li 166
Gene Symbols RNASE6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNASE6. Recombinant protein control fragment (Product #RP-88855).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6039
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.