Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ RNASE7 (Human) ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
- RNASE7 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human RNASE7.
Specifications
Specifications
Description | ribonuclease, RNase A family, 7 |
Cross Reactivity | Human |
Product Type | Kit |
Gene ID (Entrez) | 84659 |
Gene Symbol | RNASE7 |
Quantity | 1 Kit |
Storage Requirements | Store the kit at 4°C. Avoid repeated freezing and thawing. |
Target | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction