Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNASE7 Monoclonal Antibody (CL0224)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524591

Catalog No. PIMA524591


Only null left
Add to Cart

Description

Description

Immunogen sequence: KGMTSSQWFK IQHMQPSPQA CNSAMKNINK HTKRCKDLNT FLHEPFSSVA ATCQTPKIAC KNGDKNCHQS HGPVSLTMCK LTSGKYPNCR YKEKRQNKSY VVACKPPQKK DSQQFHLVPV HLDRV Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 56%. Binds to an epitope located within the peptide sequence KGMTSSQWFKIQHMQ as determined by overlapping synthetic peptides.

The RNASE7 gene encodes a ribonuclease enzyme characterized by its potent ribonucleolytic activity and antimicrobial properties. It is part of the RNase A superfamily, featuring eight conserved cysteines and a catalytic histidine-lysine-histidine triad. RNase 7 is expressed in various somatic tissues, including the liver, kidney, skeletal muscle, and heart, and is prominently found in the stratum corneum of human skin. Its role is crucial in innate immune defense, assisting in the high resistance of human skin against infections. The expression of RNASE7 can be significantly induced by pro-inflammatory cytokines such as interferon-?, interleukin 1β, and to a lesser extent by tumor necrosis factor-a. This induction highlights its role in immune responses. Beyond its antimicrobial activity, RNase 7 has been shown to play a role in translating self-nucleic acids into TLR9 activators, influencing the DNA-mediated activation of immune cells like keratinocytes. These functions underscore RNASE7's significance in both maintaining skin barrier function and modulating immune responses.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNASE7
Monoclonal
1.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H1E1
RNASE7
Recombinant protein corresponding to Human RNASE7.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
CL0224
Unconjugated
RNASE7
RAE1; Ribonuclease 7; ribonuclease A family member 7; ribonuclease, RNase A family, 7; RNase 7; RNASE7; SAP-2; skin-derived antimicrobial protein 2; UNQ2516/PRO6006
Mouse
Protein A
RUO
84659
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.