Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNASE7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552184
Description
Immunogen sequence: KGMTSSQWFK IQHMQPSPQA CNSAMKNINK HTKRCKDLNT FLHEPFSSVA ATCQTPKIAC KNGDKNCHQS HGPVSLTMCK LTSGKYPNCR YKEKRQNKSY VVACKPPQKK DSQQFHLVPV HLDRV Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 56%.

Specifications
RNASE7 | |
Polyclonal | |
Unconjugated | |
RNASE7 | |
RAE1; Ribonuclease 7; ribonuclease A family member 7; ribonuclease, RNase A family, 7; RNase 7; RNASE7; SAP-2; skin-derived antimicrobial protein 2; UNQ2516/PRO6006 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84659 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1E1 | |
RNASE7 | |
Recombinant protein corresponding to Human RNASE7. Recombinant protein control fragment (Product #RP-88942). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction