Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNASE9 Monoclonal Antibody (OTI4F12)
GREENER_CHOICE

Catalog No. PIMA527429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527429 Supplier Invitrogen™ Supplier No. MA527429
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNASE9 Monoclonal Antibody for Western Blot, IHC (P)

Does not exhibit any ribonuclease activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNASE9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNASE9
Gene Accession No. P60153
Gene Alias h461; HEL128; Inactive ribonuclease-like protein 9; RAK1; ribonuclear enzyme; ribonuclease A family member 9 (inactive); ribonuclease A K1; ribonuclease, RNase A family, 9 (non-active); RNASE9
Gene Symbols RNASE9
Host Species Mouse
Immunogen Full length human recombinant protein of RNASE9 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390443
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.