Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNASE9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549255
Description
RNASE9 Polyclonal Antibody for Western Blot
Does not exhibit any ribonuclease activity.
Specifications
RNASE9 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
P60153 | |
RNASE9 | |
KLH conjugated synthetic peptide between 139-167 amino acids from the C-terminal region of human RNASE9. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RNASE9 | |
h461; HEL128; Inactive ribonuclease-like protein 9; RAK1; ribonuclear enzyme; ribonuclease A family member 9 (inactive); ribonuclease A K1; ribonuclease, RNase A family, 9 (non-active); RNASE9 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
390443 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction