Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNaseH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530974
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530974 Supplier Invitrogen™ Supplier No. PA530974
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HCT116, whole zebrafish. Predicted reactivity: Mouse (90%), Rat (90%), Dog (94%), Pig (88%), Rabbit (91%), Sheep (88%), Rhesus Monkey (96%), Chimpanzee (100%), Bovine (88%), Guinea pig (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RNASEH1 belongs to the RNase H family. It contains 1 RNase H domain. RNASEH1 is an endonuclease that specifically degrades the RNA of RNA-DNA hybrids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNaseH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RNASEH1
Gene Accession No. O60930
Gene Alias H1RNA; LOC613354 protein; PEOB2; Ribonuclease H type II; ribonuclease H1; RNase H1; RNASEH1; RNH1; zgc:91971
Gene Symbols RNASEH1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RNase H1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246243, 436932
Target Species Human, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.