Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNaseH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560209
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560209 Supplier Thermo Scientific Supplier No. PA560209
Add to Cart
Add to Cart

RNaseH1 Polyclonal Antibody

RNaseH1 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen RNaseH1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNASEH1
Gene Accession No. O60930
Gene Alias H1RNA; LOC613354 protein; PEOB2; Ribonuclease H type II; ribonuclease H1; RNase H1; RNASEH1; RNH1; zgc:91971
Gene Symbols RNASEH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNaseH1. Recombinant protein control fragment (Product #RP-102933).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246243
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.