Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNaseH1 Recombinant Rabbit Monoclonal Antibody (HL2343)
GREENER_CHOICE

Catalog No. FEMA550496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550496 Supplier Invitrogen™ Supplier No. MA550496
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

RNaseH1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

RNASEH1 belongs to the RNase H family. It contains 1 RNase H domain. RNASEH1 is an endonuclease that specifically degrades the RNA of RNA-DNA hybrids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNaseH1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2343
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RNASEH1
Gene Accession No. O60930, O70338
Gene Alias H1RNA; LOC613354 protein; PEOB2; Ribonuclease H type II; ribonuclease H1; RNase H1; RNASEH1; RNH1; zgc:91971
Gene Symbols RNASEH1
Host Species Rabbit
Immunogen Recombinant fragment of human RNase H1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19819, 246243, 475648
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.