Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNASEH2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119399
This item is not returnable.
View return policy
Description
RNASEH2A Polyclonal Antibody for Western Blot, IHC (P)
Ribonucleases (RNAses) H are enzymes that hydrolyze the RNA strands of RNA/DNA hybrids. The major role of these enzymes is to remove the RNA strand from the RNA/DNA hybrids that form during DNA replication and repair. RNAse H2 is made up of three subunits; all three are required for RNAse activity. Recent evidence has demonstrated that mutations in RNAse H2A or any of the other subunits result in Aicardi-Goutieres syndrome (AGS), a neurological disorder with similar symptoms to viral brain infections including high levels of IFN-alpha in the cerebral spinal fluid. Similar conditions are observed with mutations in TREX1, a single-stranded DNA exonuclease, suggesting that RNAse H2 and TREX1 may have similar roles, and that mutations in any of these genes lead to an accumulation of intracellular nucleic acids, triggering an inflammatory response through activation of the innate immune system.
Specifications
RNASEH2A | |
Polyclonal | |
Unconjugated | |
RNASEH2A | |
2400006P09Rik; AGS4; Aicardi-Goutieres syndrome 4 protein; hypothetical protein LOC512830; hypothetical protein MGC56307; JUNB; ribonuclease H2 subunit A; ribonuclease H2, large subunit; ribonuclease H2, subunit A; ribonuclease HI; ribonuclease HI large subunit; Ribonuclease HI subunit A; ribonuclease HI, large subunit; RNase H(35); RNase H2 subunit A; RNase HI large subunit; Rnaseh2a; RNASEHI; RNHIA; RNHL; zgc:56307 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10535 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O75792 | |
RNASEH2A | |
Recombinant protein Ribonuclease H2 Subunit A. The antigen corresponds to amino acid range 1-299 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction