Learn More
RNASEH2A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304149AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Ribonucleases (RNAses) H are enzymes that hydrolyze the RNA strands of RNA/DNA hybrids. The major role of these enzymes is to remove the RNA strand from the RNA/DNA hybrids that form during DNA replication and repair. RNAse H2 is made up of three subunits; all three are required for RNAse activity. Recent evidence has demonstrated that mutations in RNAse H2A or any of the other subunits result in Aicardi-Goutieres syndrome (AGS), a neurological disorder with similar symptoms to viral brain infections including high levels of IFN-alpha in the cerebral spinal fluid. Similar conditions are observed with mutations in TREX1, a single-stranded DNA exonuclease, suggesting that RNAse H2 and TREX1 may have similar roles, and that mutations in any of these genes lead to an accumulation of intracellular nucleic acids, triggering an inflammatory response through activation of the innate immune system.Specifications
RNASEH2A | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O75792 | |
RNASEH2A | |
Between 250 and 300 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RNASEH2A | |
AGS4, JUNB, RNASEHI, RNHIA, RNHL; RNase H(35); RNase H2 subunit A; RNase HI large subunit; aicardi-Goutieres syndrome 4 protein; ribonuclease H2 subunit A; ribonuclease H2, large subunit; ribonuclease HI large subunit; ribonuclease HI subunit A; ribonuclease HI, large subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10535 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.