Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNASEH2B Antibody (673), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243776
Description
Ensure accurate, reproducible results in Western Blot
RNASEH2B Monoclonal specifically detects RNASEH2B in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RNASEH2B | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
RNASEH2B | |
Recombinant protein encompassing a sequence within the center region of human RNaseH2B. The exact sequence is proprietary. | |
Protein G purified | |
RUO | |
79621 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
673 | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence | |
ribonuclease H2, subunit B | |
Mouse | |
35 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction