Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNaseH2B Monoclonal Antibody (3I4)
GREENER_CHOICE

Catalog No. MA523523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA523523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA523523 Supplier Invitrogen™ Supplier No. MA523523
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: A431, HeLa, and HepG2 cells. Predicted reactivity: Pig (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNaseH2B
Applications Western Blot
Classification Monoclonal
Clone 3I4
Concentration 1.94 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RNASEH2B
Gene Accession No. Q5TBB1
Gene Alias 1110019N06Rik; 2610207P08Rik; AGS2; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; deleted in lymphocytic leukemia 8 homolog; DLEU8; ribonuclease H2; ribonuclease H2 subunit B; ribonuclease H2, subunit B; ribonuclease HI subunit B; RNase H2 subunit B; Rnaseh2b
Gene Symbols RNASEH2B
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human RNaseH2B. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79621
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.