Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNaseH2B Monoclonal Antibody (GT479)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531529
Description
Keep as concentrated solution. Predicted reactivity: Pig (85%). Positive Control: A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
Specifications
RNaseH2B | |
Monoclonal | |
1.0 mg/mL | |
PBS with 20% glycerol and no preservative; pH 7 | |
Q5TBB1 | |
RNASEH2B | |
Recombinant protein encompassing a sequence within the center region of human RNaseH2B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
GT479 | |
Unconjugated | |
RNASEH2B | |
1110019N06Rik; 2610207P08Rik; AGS2; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; deleted in lymphocytic leukemia 8 homolog; DLEU8; ribonuclease H2; ribonuclease H2 subunit B; ribonuclease H2, subunit B; ribonuclease HI subunit B; RNase H2 subunit B; Rnaseh2b | |
Mouse | |
Protein G | |
RUO | |
79621 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction