Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNaseH2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583560 Supplier Invitrogen™ Supplier No. PA583560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISDYIPKELS DDLSKYLKLP EPSASLPNPP SKKIKLSDEP VEAKEDYTKF NTKDLKTEKK NSKMTAAQKA LAKVDKSGMK SIDT Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 90%.

RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNaseH2B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNASEH2B
Gene Accession No. Q5TBB1
Gene Alias 1110019N06Rik; 2610207P08Rik; AGS2; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; deleted in lymphocytic leukemia 8 homolog; DLEU8; ribonuclease H2; ribonuclease H2 subunit B; ribonuclease H2, subunit B; ribonuclease HI subunit B; RNase H2 subunit B; Rnaseh2b
Gene Symbols RNASEH2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNASEH2B. Recombinant protein control fragment (Product #RP-106008).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79621
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.