Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNASEH2B Rabbit anti-Human, Clone: RM433, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$207.00 - $562.50

Specifications

Antigen RNASEH2B
Clone RM433
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:100-1:200
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB119710
SDP
View Documents
Novus Biologicals
NBP30918125UL
25 μg
Each for $207.00
Only null left
Add to Cart
 
NB119709
SDP
View Documents
Novus Biologicals
NBP309181100UL
100 μg
Each for $562.50
Only null left
Add to Cart
 
Description

Description

RNASEH2B Monoclonal specifically detects RNASEH2B in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

RNASEH2B
Western Blot 1:1000-1:2000, Immunohistochemistry 1:100-1:200
Monoclonal
Purified
RUO
PBS, 50% Glycerol, 1% BSA
79621
IgG
Protein A purified
RM433
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Human
ribonuclease H2, subunit B
A peptide corresponding to the C-terminus of RNASEH2B
Primary
Store at -20°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.