Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNASEH2B Recombinant Rabbit Monoclonal Antibody (RM433)
GREENER_CHOICE

Catalog No. PIMA538674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA538674 Supplier Invitrogen™ Supplier No. MA538674
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RNASEH2B Recombinant Monoclonal Antibody for Western Blot, IHC (P)

RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNASEH2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone RM433
Concentration 200 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene RNASEH2B
Gene Accession No. Q5TBB1
Gene Alias 1110019N06Rik; 2610207P08Rik; AGS2; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; deleted in lymphocytic leukemia 8 homolog; DLEU8; ribonuclease H2; ribonuclease H2 subunit B; ribonuclease H2, subunit B; ribonuclease HI subunit B; RNase H2 subunit B; Rnaseh2b
Gene Symbols RNASEH2B
Host Species Rabbit
Immunogen A peptide corresponding to the C-terminus of RNASEH2B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79621
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.