Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNaseH2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100321
Description
Antibody detects endogenous levels of total RNASEH2C.
RNH2C encodes a ribonuclease H subunit that can cleave ribonucleotides from RNA:DNA duplexes.
Specifications
RNaseH2C | |
Polyclonal | |
Unconjugated | |
RNASEH2C | |
AGS3; Aicardi-Goutieres syndrome 3 protein; AYP1; Ribonuclease H2 subunit C; ribonuclease H2, subunit C; ribonuclease HI subunit C; RNase H1 small subunit; RNase H2 subunit C; RNASEH2C | |
Rabbit | |
Affinity chromatography | |
RUO | |
84153 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8TDP1 | |
RNASEH2C | |
A synthesized peptide derived from human RNASEH2C(Accession Q8TDP1), corresponding to amino acid residues V73-T123. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction