Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNaseH2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589503
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589503 Supplier Invitrogen™ Supplier No. PA589503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MESGDEAAIE RHRVHLRSAT LRDAVPATLH LLPCEVAVDG PAPVGRFFTP AIRQGPEGLE VSFRGRCLRG EEVAVPPGLV GYVMVTEEKK VSMGKPDPLR DSGTDDQEEE PLERDFDRFI GATANFSRFT LWGLETIPGP DAKVRGALTW PSLAAAIHAQ VPED; Positive Samples: 293T, HeLa; Cellular Location: Nucleus.

RNH2C encodes a ribonuclease H subunit that can cleave ribonucleotides from RNA:DNA duplexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNaseH2C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RNASEH2C
Gene Accession No. Q8TDP1
Gene Alias AGS3; Aicardi-Goutieres syndrome 3 protein; AYP1; Ribonuclease H2 subunit C; ribonuclease H2, subunit C; ribonuclease HI subunit C; RNase H1 small subunit; RNase H2 subunit C; RNASEH2C
Gene Symbols RNASEH2C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-164 of human RNASEH2C (NP_1155692).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84153
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.