Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNaseH2C Polyclonal Antibody, Invitrogen™

Catalog No. PIPA566770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566770 Supplier Thermo Scientific Supplier No. PA566770

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: IERHRVHLRSA TLRDAVPATL HLLPCEVAVD GPAPVGRFFT PAIRQGPEGL EVSFRGRCLR GEEVAVPPGL VGYVM Highest antigen sequence identity to the following orthologs - mouse 64%, rat 66%.

RNH2C encodes a ribonuclease H subunit that can cleave ribonucleotides from RNA:DNA duplexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNaseH2C
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNASEH2C
Gene Accession No. Q8TDP1
Gene Alias AGS3; AYP1; RNase H1 small subunit; RNase H2 subunit C; aicardi-Goutieres syndrome 3 protein; ribonuclease H2 subunit C; ribonuclease HI subunit C
Gene Symbols RNASEH2C
Host Species Rabbit
Immunogen Recombit Human RNaseH2C
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84153
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.