Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNASEH2C Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501731002 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1002 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1002 Supplier Proteintech Group Inc Supplier No. 165181AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNH2C encodes a ribonuclease H subunit that can cleave ribonucleotides from RNA:DNA duplexes.

Specifications

Antigen RNASEH2C
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNASEH2C
Gene Accession No. Q8TDP1
Gene Alias AGS3, AYP1, Ribonuclease H2 subunit C, ribonuclease H2, subunit C, Ribonuclease HI subunit C, RNase H1 small subunit, RNase H2 subunit C, RNASEH2C
Gene Symbols RNASEH2C
Host Species Rabbit
Immunogen RNASEH2C Fusion Protein Ag9739
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84153
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.