Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNaseK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560799
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560799 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560799 Supplier Invitrogen™ Supplier No. PA560799
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAVLIEDVPF TEKDFENGPQ NIYNLYEQVS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

RNASEK is endoribonuclease which preferentially cleaves ApU and ApG phosphodiester bonds. RNASEK hydrolyzes UpU bonds at a lower rate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNaseK
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNASEK
Gene Accession No. Q6P5S7
Gene Alias ribonuclease K; ribonuclease kappa; ribonuclease, RNase K; RNase K; RNase kappa; RNase kappa-02 isoform; RNASEK
Gene Symbols RNASEK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNaseK. Recombinant protein control fragment (Product #RP-100358).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.