Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNaseL Recombinant Rabbit Monoclonal Antibody (HL2341)
GREENER_CHOICE

Catalog No. FEMA550495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550495 Supplier Invitrogen™ Supplier No. MA550495
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

RNaseL Recombinant Monoclonal Antibody for Western Blot

A virally induced endoribonuclease, RNase L is the terminal factor of the anti-viral action of interferon. Activation of RNase L results in inhibition of viral proliferation. Activation of RNase L has also been determined to degrade RNA, thus, initiating a cellular stress response, inducing apoptosis. Since mutations in RNase L relate to an increased incidence of prostate cancer, the activation of RNase L functions in counteracting prostate cancer, via apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNaseL
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL2341
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RNASEL
Gene Accession No. Q05823, Q05921
Gene Alias 2',5'-oligoisoadenylate synthetase-dependent; 2-5A-dependent ribonuclease; 2-5A-dependent RNAase; 2-5A-dependent RNase; E230029I04Rik; I79_009550; interferon-induced 2-5A-dependent RNase; PRCA1; Ribonuclease 4; ribonuclease L; ribonuclease L (2 5-oligoisoadenylate synthetase-dependent); ribonuclease L (2', 5'-oligoisoadenylate synthetase-dependent); ribonuclease L (2, 5-oligoisoadenylate synthetase-dependent); ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent); RNase L; Rnasel; RNS4
Gene Symbols RNASEL
Host Species Rabbit
Immunogen Recombinant fragment of human RNase L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24014, 359726, 490310, 6041
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.