Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RND1 Antibody (OTI1C3), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273919UV
Description
RND1 Monoclonal specifically detects RND1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
RND1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RND1 | |
Full length recombinant protein of human RND1 (NP_055285) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1C3 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
ARHS, GTP-binding protein, ras homolog gene family, member S, Rho family GTPase 1FLJ42294, Rho6, rho-related GTP-binding protein Rho6, RHOS, Rnd1 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
27289.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction