Learn More
RND1 Mouse anti-Human, Clone: OTI1C3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501691

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Lacks intrinsic GTPase activity. Has a low affinity for GDP, and constitutively binds GTP. Controls rearrangements of the actin cytoskeleton. Induces the Rac-dependent neuritic process formation in part by disruption of the cortical actin filaments. Causes the formation of many neuritic processes from the cell body with disruption of the cortical actin filaments.Specifications
RND1 | |
Monoclonal | |
Unconjugated | |
RND1 | |
ARHS, RHO6, RHOS | |
Mouse | |
Affinity Chromatography | |
RUO | |
27289 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1C3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q92730 | |
RND1 | |
Full length human recombit protein of human RND1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.