Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567468 Supplier Invitrogen™ Supplier No. PA567468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDLRTDLSTLM ELSHQKQAPI SYEQGCAIAK QLGAEIYLEG SAFTSEKSIH SI Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

Lacks intrinsic GTPase activity. Has a low affinity for GDP, and constitutively binds GTP. Controls rearrangements of the actin cytoskeleton. Induces the Rac-dependent neuritic process formation in part by disruption of the cortical actin filaments. Causes the formation of many neuritic processes from the cell body with disruption of the cortical actin filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RND1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rnd1
Gene Accession No. Q92730
Gene Alias A830014L09Rik; Arhs; GTP-binding protein RHO6; ras homolog gene family, member S; Rho family GTPase 1; Rho6; Rho-related GTP-binding protein Rho6; RHOS; RND1; similar to Rho-related GTP-binding protein Rho6 (Rho family GTPase 1) (Rnd1)
Gene Symbols Rnd1
Host Species Rabbit
Immunogen Recombinant Human RND1. Recombinant protein control fragment (Product #RP-108124).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27289
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.