Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RND1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589374
Description
Positive Samples: Mouse brain, Mouse liver, Rat brain; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic side, Lipid-anchor, cytoskeleton Immunogen sequence: CLNKPSPLPQ KSPVRSLSKR.
Lacks intrinsic GTPase activity. Has a low affinity for GDP, and constitutively binds GTP. Controls rearrangements of the actin cytoskeleton. Induces the Rac-dependent neuritic process formation in part by disruption of the cortical actin filaments. Causes the formation of many neuritic processes from the cell body with disruption of the cortical actin filaments.
Specifications
RND1 | |
Polyclonal | |
Unconjugated | |
Rnd1 | |
A830014L09Rik; Arhs; GTP-binding protein RHO6; ras homolog gene family, member S; Rho family GTPase 1; Rho6; Rho-related GTP-binding protein Rho6; RHOS; RND1; similar to Rho-related GTP-binding protein Rho6 (Rho family GTPase 1) (Rnd1) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
223881, 27289, 362993 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.44 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8BLR7, Q92730 | |
Rnd1 | |
A synthetic peptide corresponding to a sequence within amino acids 133-232 of human RND1 (NP_0552851). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction