Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RND3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298843MFV500
Description
RND3 Polyclonal antibody specifically detects RND3 in Human samples. It is validated for Western BlotSpecifications
RND3 | |
Polyclonal | |
Western Blot | |
ARHE, Protein MemB, Rho family GTPase 3member E, RHO8, RHOE, Rho-related GTP-binding protein Rho8, rho-related GTP-binding protein RhoE, Rnd3, small GTP binding protein Rho8 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RND3. | |
0.1 mL | |
Signal Transduction | |
390 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction