Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540866 Supplier Invitrogen™ Supplier No. PA540866
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PPRSASAGPA GESKPKSDGK NSSGSKRYNR KRELSYPKNE SFNNQSRRSS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

The protein encoded by this gene contains a ring finger motif, which is known to be involved in protein-protein interactions. The specific function of this protein has not yet been determined. EST data suggests the existence of multiple alternatively spliced transcript variants, however, their full length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF10
Gene Accession No. Q8N5U6
Gene Alias AA675014; Kiaa0262; LOW QUALITY PROTEIN: RING finger protein 10; MGC126758; MGC126764; mKIAA0262; RIE2; RING finger protein 10; Rnf10; Sid 2705; Sid2705
Gene Symbols RNF10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RNF10 (aa 36-85).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9921
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.