Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588469
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588469 Supplier Invitrogen™ Supplier No. PA588469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SMSEDVRQRH RYLSHLPLTC EFSICELALQ PPVVSKETLE MFSDDIEKRK RQRQKKAREE RRRERRIEIE ENKKQGKYPE VHIPLENLQQ FPAFNSYTCS SDSALGPTST EGHGALSISP LSRSPGSHAD FLLTPLSPTA SQGSPSFCVG SLEEDSPFPS FAQMLRVGKA KADVWPKTAP KKDENSLVPP APVDSDGESD NSDRVPVPSF QNSFSQAIEA AFMKLDTPAT SDPLSEEKGG KKRKKQKQKL LFSTSVVHTK; Positive Samples: Raji, Mouse heart, Mouse brain; Cellular Location: Cytoplasm, Nucleus.

The protein encoded by this gene contains a ring finger motif, which is known to be involved in protein-protein interactions. The specific function of this protein has not yet been determined. EST data suggests the existence of multiple alternatively spliced transcript variants, however, their full length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene RNF10
Gene Accession No. Q5XI59
Gene Alias AA675014; Kiaa0262; LOW QUALITY PROTEIN: RING finger protein 10; MGC126758; MGC126764; mKIAA0262; RIE2; RING finger protein 10; Rnf10; Sid 2705; Sid2705
Gene Symbols RNF10
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288710
Target Species Rat, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.