Learn More
RNF10 Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human RNF10.
Supplier: Abnova Corporation PAB29448
Description
The protein encoded by this gene contains a ring finger motif, which is known to be involved in protein-protein interactions. The specific function of this protein has not yet been determined. EST data suggests the existence of multiple alternatively spliced transcript variants, however, their full length nature is not known. [provided by RefSeq]
Sequence: SPCYYFYQAEDGQHMFLHPVNVRCLVREYGSLERSPEKISATVVEIAGYSMSEDVRQRHRYLSHLPLTCEFSICELALQPPVVSpecifications
RNF10 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human RNF10. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
RNF10 | |
Recombinant protein corresponding to human RNF10. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
KIAA0262/MGC126758/MGC126764/RIE2 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
9921 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.