Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF11 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298851AF594
Description
RNF11 Polyclonal specifically detects RNF11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RNF11 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CGI-123, MGC51169, ring finger protein 11, SID1669, Sid1669p | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human RNF11. | |
0.1 mL | |
Zinc Finger | |
26994 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction