Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF11 Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
SDP

Catalog No. 502543981
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3981 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3981 Supplier OriGene Supplier No. CF810976
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNF11 Monoclonal antibody specifically detects RNF11 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen RNF11
Applications Western Blot
Classification Monoclonal
Clone OTI1G7
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene IMPDH2
Gene Accession No. E9PU28, P12268, P24547
Gene Alias CGI-123; N4wbp2; NEDD4 WW domain-binding protein 2; RING finger protein 11; Rnf11; Sid 1669; SID1669
Host Species Mouse
Immunogen Full length human recombinant protein of human IMPDH2 (NP_000875) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23918, 301005, 3615
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.