Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF111 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558449 Supplier Invitrogen™ Supplier No. PA558449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTPSDEDNDS SFSDCLSSPS SSLHFGDSDT VTSDEDKEVS VRHSQTILNA KSRSHSARSH KWPRTETESV SGLLMKRPCL HGSSLRRLPC Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 86%.

The protein encoded by this gene is a nuclear RING-domain containing E3 ubiquitin ligase. This protein interacts with the transforming growth factor (TGF) -beta/NODAL signaling pathway by promoting the ubiquitination and proteosomal degradation of negative regulators, like SMAD proteins, and thereby enhances TGF-beta target-gene transcription. As a modulator of the nodal signaling cascade, this gene plays a critical role in the induction of mesoderm during embryonic development. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Jul 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF111
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF111
Gene Accession No. Q6ZNA4, Q99ML9
Gene Alias ARK; Arkadia; E3 ubiquitin-protein ligase Arkadia; hRNF111; ring finger 111; RING finger protein 111; RING-type E3 ubiquitin transferase Arkadia; Rnf111
Gene Symbols RNF111
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF111. Recombinant protein control fragment (Product #RP-98114).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54778, 93836
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.