Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF113A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585006 Supplier Invitrogen™ Supplier No. PA585006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAKELIAKLE KHRATGEGGA SDLPEDPDED AIPIT Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 66%.

This intronless gene encodes a protein which contains a C3H1-type zinc finger domain and a C3HC4 Ring-type (Really Interesting New Gene-type) zinc finger domain. The Ring-type zinc finger domain is identified in various tumor suppressors, DNA repair genes and cytokine receptor-associated molecules, and is probably involved in mediating protein-protein interactions. [provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF113A
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF113A
Gene Accession No. O15541
Gene Alias 2810428C21Rik; Cwc24; Cwc24 homolog; E3 ubiquitin-protein ligase RNF113A; RING finger protein 113A; ring finger protein 113A1; RNF113; RNF113A; Rnf113a1; TTD5; Zinc finger protein 183; zinc finger protein 183 (RING finger, C3HC4 type); ZNF183
Gene Symbols RNF113A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF113A. Recombinant protein control fragment (Product #RP-108669).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7737
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.