Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF113A Rabbit anti-Human, Polyclonal, ProteinTech
SDP

Catalog No. 502238548 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8548 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-8548 Supplier Proteintech Group Inc Supplier No. 270181AP150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aliquoting is unnecessary for -20°C storage.

RNF113A Polyclonal antibody specifically detects RNF113A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RNF113A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 333 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene RNF113A
Gene Accession No. O15541
Gene Alias 2810428C21Rik; Cwc24; Cwc24 homolog; E3 ubiquitin-protein ligase RNF113A; RING finger protein 113A; ring finger protein 113A1; RNF113; RNF113A; Rnf113a1; TTD5; Zinc finger protein 183; zinc finger protein 183 (RING finger, C3HC4 type); ZNF183
Gene Symbols RNF113A
Host Species Rabbit
Immunogen RNF113A Fusion Protein Ag25313
Purification Method Antigen affinity chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7737
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.