Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF113B Monoclonal Antibody (OTI2A6), TrueMAB™, OriGene
SDP

Catalog No. 501674873
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4873 100 μL
50-254-0501 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-4873 Supplier OriGene Supplier No. TA504119
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNF113B (Ring Finger Protein 113B) is a Protein Coding gene. An important paralog of this gene is RNF113A.

Specifications

Antigen RNF113B
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2A6
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RNF113B
Gene Accession No. Q8IZP6
Gene Alias RNF161, ZNF183L1, bA10G5.1
Gene Symbols RNF113B
Host Species Mouse
Immunogen Full length human recombit protein of human RNF113B produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140432
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.