Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF113B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113600
Description
Immunogen sequence: MGATADFEQD TEKEHHTPTI LKCSQRVQEA LRGREHDHIY RGIHSYLRYL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
RNF113B (Ring Finger Protein 113B) is a Protein Coding gene. An important paralog of this gene is RNF113A.
Specifications
RNF113B | |
Polyclonal | |
Unconjugated | |
RNF113B | |
bA10G5.1; RING finger protein 113B; RNF113B; RNF161; Zinc finger protein 183-like 1; ZNF183L1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
140432 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IZP6 | |
RNF113B | |
Synthetic peptide directed towards the middle region of human RNF113B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction