Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF113B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988945
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8945 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8945 Supplier Bioss Supplier No. BS9256R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF113B (Ring Finger Protein 113B) is a Protein Coding gene. An important paralog of this gene is RNF113A.

Specifications

Antigen RNF113B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RNF113B
Gene Accession No. Q8IZP6
Gene Alias bA10G5.1; RING finger protein 113B; RNF113B; RNF161; Zinc finger protein 183-like 1; ZNF183L1
Gene Symbols RNF113B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RNF113B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140432
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.