Learn More
Invitrogen™ RNF12 Monoclonal Antibody (OTI2D7)

Description
RNF12 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RNF12 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2D7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RLIM |
| Gene Accession No. | Q9NVW2 |
| Gene Alias | AL022832; AW743871; CTD-2530H13.3; DKFZp686N06224; E3 ubiquitin-protein ligase RLIM; E3 ubiquitin-protein ligase RNF12; FLJ25923; FLJ41093; FLJ42887; FLJ45986; Ha1r; Hoxa1 regulated; LIM domain-interacting RING finger protein; MGC15161; NY-REN-43; renal carcinoma antigen NY-REN-43; RING finger LIM domain-binding protein; ring finger protein 12; ring finger protein, LIM domain interacting; ring zinc finger LIM domain binding protein; ring zinc finger protein NY-REN-43antigen; RING-type E3 ubiquitin transferase RLIM; Rlim; R-LIM; Rlim1; RNF12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.