Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529530 Supplier Invitrogen™ Supplier No. PA529530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, Raji, K562. Predicted reactivity: Rat (80%), Rhesus Monkey (99%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RNF12 is a RING-H2 zinc finger protein. It has been shown to be a ubiquitin protein ligase that targets LIM domain binding 1 (LDB1/CLIM), and causes proteasome-dependent degradation of LDB1. This protein and LDB1 are co-repressors of LHX1/LIM-1, a homeodomain transcription factor. Alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF12
Applications Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene RLIM
Gene Accession No. Q9NVW2
Gene Alias AL022832; AW743871; CTD-2530H13.3; DKFZp686N06224; E3 ubiquitin-protein ligase RLIM; E3 ubiquitin-protein ligase RNF12; FLJ25923; FLJ41093; FLJ42887; FLJ45986; Ha1r; Hoxa1 regulated; LIM domain-interacting RING finger protein; MGC15161; NY-REN-43; renal carcinoma antigen NY-REN-43; RING finger LIM domain-binding protein; ring finger protein 12; ring finger protein, LIM domain interacting; ring zinc finger LIM domain binding protein; ring zinc finger protein NY-REN-43antigen; RING-type E3 ubiquitin transferase RLIM; Rlim; R-LIM; Rlim1; RNF12
Gene Symbols RLIM
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RNF12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51132
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.