Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF121 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561136 Supplier Invitrogen™ Supplier No. PA561136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAVVEVEVG GGAAGERELD EVDMSDLSPE EQWRVEHARM HAKHRGHEAM HA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Several alternatively spliced transcript variants have been noted for this gene, however, not all are likely to encode viable protein products.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF121
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF121
Gene Accession No. Q9H920
Gene Alias 4930544L10Rik; RING finger protein 121; Rnf121
Gene Symbols RNF121
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF121. Recombinant protein control fragment (Product #RP-100551).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55298
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.