Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF122 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552013 Supplier Invitrogen™ Supplier No. PA552013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NQAQSERYGY KEVVLKGDAK KLQLYGQTCA VCLEDFKGKD ELGVLPCQHA FHRKCLVKWL EVRCVCPMCN KPIASPSEAT QNIGILLDEL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

May induce necrosis and apoptosis. May play a role in cell viability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF122
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF122
Gene Accession No. Q9H9V4
Gene Alias 1110063C11Rik; FLJ12526; MGC126622; RGD1561238; RING finger protein 122; ring finger protein 122 homolog; RN122; RNF122
Gene Symbols RNF122
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF122. Recombinant protein control fragment (Product #RP-102195).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79845
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.