Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF122 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113768 Supplier Invitrogen™ Supplier No. PA5113768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLIFCCYFIS KLRNQAQSER YGYKEVVLKG DAKKLQLYGT CAVCLEDFKG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

May induce necrosis and apoptosis. May play a role in cell viability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF122
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF122
Gene Accession No. Q8BP31
Gene Alias 1110063C11Rik; FLJ12526; MGC126622; RGD1561238; RING finger protein 122; ring finger protein 122 homolog; RN122; RNF122
Gene Symbols RNF122
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of Mouse Rnf122 (aa 52-102).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68867
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.