Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF123 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RNF123 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RNF123 Polyclonal specifically detects RNF123 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RNF123 | |
Polyclonal | |
Rabbit | |
Zinc Finger | |
DKFZp686C2222, E3 ubiquitin-protein ligase RNF123, EC 6.3.2.-, FLJ12565, Kip1 ubiquitination-promoting complex protein 1, KPC1, ring finger protein 123MGC163504, ubiquitin ligase | |
RNF123 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
63891 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LSTSEKVKVRTLSVEQRTREDIEGSHWNEGLLLGRPPEEPEQPLTENSLLEVLDGAVMMYNLSVHQQLGKMVGVSDDVNEYAMA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title