Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF123 Mouse anti-Human, Clone: 2C10.1, MilliporeSigma™

Catalog No. MABS1896100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1896-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1896-100 Supplier MilliporeSigma Supplier No. MABS1896100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RNF123, clone 2C10.1 is a mouse monoclonal antibody that detects RING-type E3 ubiquitin transferase RNF123 and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF123
Applications Western Blot
Classification Monoclonal
Clone 2C10.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols RNF123,KPC1,PF1477
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 92 amino acids from the N-terminal region of human RING-type E3 ubiquitin transferase RNF123.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_071347
Test Specificity Clone 2C10.1 specifically detects RING-type E3 ubiquitin transferase RNF123 in human cells. It targets an epitope within 92 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.