Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF123 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568183 Supplier Invitrogen™ Supplier No. PA568183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

Catalytic subunit of the KPC complex that acts as E3 ubiquitin-protein ligase. Required for poly-ubiquitination and proteasome-mediated degradation of CDKN1B during G1 phase of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF123
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RNF123
Gene Accession No. Q5XPI3, Q5XPI4
Gene Alias BC003945; E3 ubiquitin-protein ligase RNF123; FP1477; Kip1 ubiquitination-promoting complex protein 1; KPC1; RING finger protein 123; RING-type E3 ubiquitin transferase RNF123; RNF123; ubiquitin ligase
Gene Symbols RNF123
Host Species Rabbit
Immunogen Synthesized peptide derived from human RNF123.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63891, 84585
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.